missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UTP15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15878893
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15878893 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15878893 Supplier Invitrogen™ Supplier No. PA570487

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UTP15
Gene Accession No. Q8C7V3
Gene Alias AW544865; LOW QUALITY PROTEIN: U3 small nucleolar RNA-associated protein 15 homolog; NET21; RGD1310992; Src-associated protein SAW; U3 small nucleolar RNA-associated protein 15 homolog; U3 small nucleolar RNA-associated protein 15 homolog; LOW QUALITY PROTEIN: U3 small nucleolar RNA-associated protein 15 homolog; UniProtKB:Q8TED0}; UTP15; utp15 {ECO:0000250; UTP15 small subunit processome component; UTP15 SSU processome component; UTP15, small subunit processome component; UTP15, U3 small nucleolar ribonucleoprotein, homolog; UTP15, U3 small nucleolar ribonucleoprotein, homolog (S. cerevisiae); UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast)
Gene Symbols UTP15
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human UTP15 (aa 301-350).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105372
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.