missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTP14C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562899
This item is not returnable.
View return policy
Description
Immunogen sequence: RVQTLEELEE LGKEDCFQNK ELPRPVLEGQ QSERTPNNRP DAPKEKKEKE Q Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 57%.
Specifications
| UTP14C | |
| Polyclonal | |
| Unconjugated | |
| UTP14C | |
| 2700066J21Rik; KIAA0266; U3 small nucleolar RNA-associated protein 14 homolog C; UTP14, small subunit processome component homolog C (S. cerevisiae); UTP14, U3 small nucleolar ribonucleoprotein, homolog C; UTP14B; UTP14C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9724 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TAP6 | |
| UTP14C | |
| Recombinant protein corresponding to Human UTP14C. Recombinant protein control fragment (Product #RP-99595). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction