missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP14A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538778
This item is not returnable.
View return policy
Description
This gene encodes a member of the uridine triphosphate 14 family. As an essential component of a large ribonucleoprotein complex bound to the U3 small nucleolar RNA, the encoded protein is involved in ribosome biogenesis and 18S rRNA synthesis. An autosomal retrotransposed copy of this X-linked gene exists on chromosome 13. Alternative splicing results in multiple transcript variants.[provided by RefSeq, Oct 2009].
Specifications
| UTP14A | |
| Polyclonal | |
| Unconjugated | |
| UTP14A | |
| NYCO16, SDCCAG16, dJ537K23.3, U3 small nucleolar RNA-associated protein 14 homolog A, antigen NY-CO-16, serologically defined colon cancer antigen 16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10813, 72554 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BVJ6, Q640M1 | |
| UTP14A | |
| A synthetic peptide derived from the internal region of human UTP14A | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction