missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP14A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93627-0.02ml
This item is not returnable.
View return policy
Description
UTP14A Polyclonal antibody specifically detects UTP14A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| UTP14A | |
| Polyclonal | |
| Western Blot 1:200-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Antigen NY-CO-16, dJ537K23.3, FLJ37089, NYCO16, NY-CO-16, SDCCAG16, Serologically defined colon cancer antigen 16KIAA0266, U3 small nucleolar RNA-associated protein 14 homolog A, UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human UTP14A (NP_006640.2). MTANRLAESLLALSQQEELADLPKDYLLSESEDEGDNDGERKHQKLLEAISSLDGKNRRKLAERSEASLKVSEFNVSSEGSGEKLVLADLLEPVKTSSSLATVKKQLSRVKSKKTVELPLNKEEIERIHREVAFNKTAQVLSKWDPVVLKNRQAEQLVFPLEKEEPAIAPIEHVLSGWKARTPLEQEIFNLLHKNKQPVTDPLLTPVEKASLRAMSLEEAKMRRAELQRARALQSYYEAKARREKKIKSKKYHKVVKKGK | |
| 0.02 mL | |
| Cancer, Immunology | |
| 10813 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction