missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTP11L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116839
This item is not returnable.
View return policy
Description
Positive Samples: Mouse testis, Mouse stomach, Rat spleen Immunogen sequence: VAEAKKIERL KSELHLLDFQ GKQQNKHVFF FDTKKEVEQF DVATHLQTAP ELVDRVFNRP RIETLQKEKV KGVTNQTGLK RIAKERQKQY NCLTQRIERE KKLFVIAQKI QTRKDLMDKT QKVKVKKETV N.
Involved in nucleolar processing of pre-18S ribosomal RNA. Target protein may have a role in maintainance of neuronal function.
Specifications
| UTP11L | |
| Polyclonal | |
| Unconjugated | |
| UTP11 | |
| 2700082D03Rik; Cgi94; CGI-94; HDCMB12P; Probable U3 small nucleolar RNA-associated protein 11; RGD1559610; U3 snoRNA-associated protein 11; UTP11; UTP11, small subunit processome component homolog (S. cerevisiae); UTP11L; UTP11-like protein; UTP11-like, U3 small nucleolar ribonucleoprotein; UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 313581, 51118, 67205 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.65 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8R5K5, Q9CZJ1, Q9Y3A2 | |
| UTP11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-240 of human UTP11L (NP_0571212). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido