missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540319
This item is not returnable.
View return policy
Description
Peptide sequence: AREDPDSPPG RPEDCAPPPA APPSLNTALL QTLGHLGDIA NILGPLRDQL Sequence homology: Cow: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 86%.
Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.
Specifications
| UTF1 | |
| Polyclonal | |
| Unconjugated | |
| UTF1 | |
| AI505934; OTTHUMP00000020786; Undifferentiated embryonic cell transcription factor 1; UTF1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8433 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5T230 | |
| UTF1 | |
| Synthetic peptide directed towards the middle region of human UTF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction