missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTF1 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149407
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.
Specifications
| UTF1 | |
| Polyclonal | |
| Unconjugated | |
| UTF1 | |
| AI505934; OTTHUMP00000020786; Undifferentiated embryonic cell transcription factor 1; UTF1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 22286 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6J1H4 | |
| UTF1 | |
| E. coli-derived recombinant mouse UTF1, Lys50-Ser171. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction