missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12788-100ul
This item is not returnable.
View return policy
Description
UTF1 Polyclonal antibody specifically detects UTF1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| UTF1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
| undifferentiated embryonic cell transcription factor 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human UTF1 | |
| 100 μg | |
| Stem Cell Markers | |
| 8433 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction