missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uteroglobin/SCGB1A1 Antibody (21B2), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Uteroglobin/SCGB1A1 |
|---|---|
| Clone | 21B2 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30026125
|
Novus Biologicals
NBP3-26335-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30038510
|
Novus Biologicals
NBP3-26335-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Uteroglobin/SCGB1A1 Monoclonal antibody specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Uteroglobin/SCGB1A1 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
| A synthesized peptide derived from Human Uteroglobin/SCGB1A1 [UniProt P11684] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 21B2 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cytokine Research | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 7356 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title