missing translation for 'onlineSavingsMsg'
Learn More

Uteroglobin/SCGB1A1 Antibody (1F7), Novus Biologicals™

Product Code. 18375307 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375307 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375307 Supplier Novus Biologicals Supplier No. H00007356M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Uteroglobin/SCGB1A1 Monoclonal antibody specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Uteroglobin/SCGB1A1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1F7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH04481
Gene Alias blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin
Host Species Mouse
Immunogen SCGB1A1 (AAH04481, 22 a.a. ~ 91 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EICPSFQRVIETLLMDTPSSYEAAMELFSPDQDMREAGAQLKKLVDTLPQKPRESIIKLMEKIAQSSLCN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 7356
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.