Learn More
PeproTech Uteroglobin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P330BT-25UG
Description
AA Sequence of recombinant protein: MEICPSFQRV IETLLMDTPS SYEAAMELFS PDQDMREAGA QLKKLVDTLP QKPRESIIKL MEKIAQSSLC N. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Uteroglobin. Anti-Human Uteroglobin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Uteroglobin by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Uteroglobin (500-P330) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human Uteroglobin. Western Blot: To detect Human Uteroglobin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human Uteroglobin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P330BT-1MG will be provided as 2 x 500 μg
Uteroglobin, which is a member of the Secretoglobin superfamily and is also known as Clara cell phospholipid-binding protein, is a multifunctional protein that can exert anti-inflammatory and anti-tumorigenic effects by binding small hydrophobic molecules such as phospholipids and prostaglandins. The small, non-glycosylated protein named for its high levels of expression in pre-implantation embryos, where it exhibits growth stimulatory effects, is produced and secreted by the non-ciliated, non-mucous Clara cells predominant in the epithelial surfaces of pulmonary airways, as well as other non-ciliated epithelia. Members of the Secretoglobin superfamily demonstrate a high level of structural conservation and are characterized as small, secretory homo- or heterodimers. In addition to sequestering pro-inflammatory mediators and carcinogens, Uteroglobin has been implicated in the inhibition of cell migration and invasion, platelet aggregation, and T cell differentiation.
Specifications
| Uteroglobin | |
| Polyclonal | |
| Biotin | |
| SCGB1A1 | |
| Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell 17 kDa protein; clara cell phospholipid-binding protein; clara cell secretory protein; clara cells 10 kDa secretory protein; PCBB; PCB-binding protein; PCB-BP; SCGB1A1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UG; Ugb; UP1; UP-1; Urinary protein 1; urine protein 1; uteroglobin; Uteroglobin (Clara cell secretory protein); Utg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7356 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P11684 | |
| SCGB1A1 | |
| E.coli-derived Recombinant Human Uteroglobin. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.