missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UT2/SLC14A2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-44375CL1
This item is not returnable.
View return policy
Description
UT2/SLC14A2 Polyclonal antibody specifically detects UT2/SLC14A2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| UT2/SLC14A2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| FLJ16167, HUT2UT-A2, hUT-A6, MGC119566, solute carrier family 14 (urea transporter), member 2, Solute carrier family 14 member 2, urea transporter 2, Urea transporter, kidney, urea transporter-2, UT2MGC119567, UTA, UTR | |
| Produced against a synthetic peptide mapped to the C-terminal tail (amino acids 911-929) of rat UT-A1 (antibody designation L194) | |
| 0.1 mL | |
| ABC Transporters, Neuroscience | |
| 8170 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction