missing translation for 'onlineSavingsMsg'
Learn More

UT2/SLC14A2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£155.00 - £364.00

Specifications

Antigen UT2/SLC14A2
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18606671
View Documents
Novus Biologicals
NBP2-93774-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18629920
View Documents
Novus Biologicals
NBP2-93774-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

UT2/SLC14A2 Polyclonal antibody specifically detects UT2/SLC14A2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

UT2/SLC14A2
Western Blot
Unconjugated
Rabbit
ABC Transporters, Neuroscience
PBS (pH 7.3), 50% glycerol
8170
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human, Mouse
FLJ16167, HUT2UT-A2, hUT-A6, MGC119566, solute carrier family 14 (urea transporter), member 2, Solute carrier family 14 member 2, urea transporter 2, Urea transporter, kidney, urea transporter-2, UT2MGC119567, UTA, UTR
Recombinant fusion protein containing a sequence corresponding to amino acids 420-580 of human SLC14A2 (NP_009094.3). FRLPLSKVTYPEANRIYYLTVKSGEEEKAPSGGGGEHPPTAGPKVEEGSEAVLSKHRSVFHIEWSSIRRRSKVFGKGEHQERQNKDPFPYRYRKPTVELLDLDTMEESSEIKVETNISKTSWIRSSMAASGKRVSKALSYITGEMKECGEGLKDKSPVFQF
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.