missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody has been tested in direct-ELISA.
RNase U2 is a 114aa endoribonuclease secreted by the fungus Ustilago sphaerogena. RNase U2 displays optimum pH for enzymatic activity at pH 3.5-5.5.
Specifications
Specifications
| Antigen | Ustilago sphaerogena RNU2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 3.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | Ribonuclease U2; RNase U2 |
| Host Species | Rabbit |
| Immunogen | Recombinant Ustilago sphaerogena Ribonuclease U2 protein (1-114aa). |
| Purification Method | Protein G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
