missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP9X Recombinant Rabbit Monoclonal Antibody (23GB2000), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552470
This item is not returnable.
View return policy
Description
USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.
Specifications
| USP9X | |
| Recombinant Monoclonal | |
| Unconjugated | |
| USP9X | |
| 5730589N07Rik; AA407302; AA407699; AL022658; AL022749; Deubiquitinating enzyme FAF-X; DFFRX; Drosophila fat facets related, X-linked; FAF; Fafl; FAF-X; FAM; fat facets homolog; Fat facets in mammals; fat facets protein related, X-linked; fat facets protein-related, X-linked; fats facets protein related, X; hFAM; MRX99; MRXS99F; Probable ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-x-like protein; RGD1560056; Ubiquitin carboxyl-terminal hydrolase FAM; ubiquitin specific peptidase 9 X-linked; ubiquitin specific peptidase 9, X chromosome; ubiquitin specific peptidase 9, X-linked; ubiquitin specific protease 9 SSSRF- isoform; ubiquitin specific protease 9, X chromosome; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); ubiquitin specific protease 9, X-linked; ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X; ubiquitin-specific protease 9, X chromosome; Ubiquitin-specific-processing protease FAF-X; USP9; USP9X | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22284, 363445, 8239 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2000 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70398, Q93008 | |
| USP9X | |
| A synthesized peptide derived from human USP9x (1-58AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction