missing translation for 'onlineSavingsMsg'
Learn More

USP9x Antibody (OTI2G7), DyLight 594, Novus Biologicals™

Product Code. 18423868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423868 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423868 Supplier Novus Biologicals Supplier No. NBP274822DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP9x Monoclonal antibody specifically detects USP9x in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP9x
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2G7
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme FAF-X, DFFRXubiquitin specific protease 9, X-linked (fat facets-like, Drosophila), Drosophila fat facets related, X-linked, EC 3.1.2.15, EC 3.4.19.12, FAF, FAM, Fat facets in mammals, fat facets protein related, X-linked, Fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific peptidase 9, X-linked, ubiquitin specific peptidase 9, X-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-X, Ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, Ubiquitin-specific protease 9, X chromosome, Ubiquitin-specific-processing protease FAF-X, USP9, X chromosome (fat facets-like Drosophila)
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2246-2570 of human USP9x (NP_001034680) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8239
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.