missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP7 Rabbit anti-Human, Mouse, Rat, Clone: 3J7G1, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | USP7 |
|---|---|
| Clone | 3J7G1 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18357484
|
Novus Biologicals
NBP3-16188-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342293
|
Novus Biologicals
NBP3-16188-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP7 Monoclonal antibody specifically detects USP7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| USP7 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Deubiquitinating enzyme 7, EC 3.1.2.15, EC 3.4.19.12, HAUSPTEF1, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, ubiquitin carboxyl-terminal hydrolase 7, ubiquitin specific peptidase 7 (herpes virus-associated), ubiquitin specific protease 7 (herpes virus-associated), Ubiquitin thioesterase 7, Ubiquitin-specific-processing protease 7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-200 of human USP7 (Q93009). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3J7G1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, DNA Repair, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 7874 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title