missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP7 Antibody (5F11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51598-0.025ml
This item is not returnable.
View return policy
Description
USP7 Monoclonal antibody specifically detects USP7 in Human samples. It is validated for Western Blot, ELISA
Specifications
| USP7 | |
| Monoclonal | |
| Unconjugated | |
| Ascites | |
| Deubiquitinating enzyme 7, EC 3.1.2.15, EC 3.4.19.12, HAUSPTEF1, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, ubiquitin carboxyl-terminal hydrolase 7, ubiquitin specific peptidase 7 (herpes virus-associated), ubiquitin specific protease 7 (herpes virus-associated), Ubiquitin thioesterase 7, Ubiquitin-specific-processing protease 7 | |
| Purified recombinant fragment of human USP7 expressed in E. Coli. | |
| 0.025 mL | |
| Apoptosis, Cell Biology, DNA Repair, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
| 7874 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 5F11 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Q93009 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction