missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP6NL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47264
This item is not returnable.
View return policy
Description
USP6NL Polyclonal specifically detects USP6NL in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).
Specifications
| USP6NL | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| USP6NL | |
| The immunogen for this product maps to a region between residue 350 and 400 of human Related to the N Terminus of tre using the numbering given in entry AAH42943.1 (GeneID 9712). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation (Negative) | |
| KIAA0019RNTRErelated to the N terminus of tre, Related to the N-terminus of tre, RN-tre, TRE2NL, USP6 N-terminal like, USP6 N-terminal-like protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9712 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction