missing translation for 'onlineSavingsMsg'
Learn More

USP6NL Antibody, DyLight 550, Novus Biologicals™

Product Code. 18410126 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410126 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410126 Supplier Novus Biologicals Supplier No. NBP147264R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP6NL Polyclonal antibody specifically detects USP6NL in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP6NL
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0019RNTRErelated to the N terminus of tre, Related to the N-terminus of tre, RN-tre, TRE2NL, USP6 N-terminal like, USP6 N-terminal-like protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 350 and 400 of human Related to the N Terminus of tre using the numbering given in entry AAH42943.1 (GeneID 9712).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9712
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.