missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP54 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103905
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP54.
USP54 has no peptidase activity.
Specifications
| USP54 | |
| Polyclonal | |
| Unconjugated | |
| USP54 | |
| 4930429G18Rik; AI115571; bA137L10.3; bA137L10.4; C030002J06Rik; C10orf29; Inactive ubiquitin carboxyl-terminal hydrolase 54; inactive ubiquitin-specific peptidase 54; ubiquitin specific peptidase 54; ubiquitin specific protease 54; ubiquitin specific proteinase 54; Usp54 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 159195, 408223, 78787 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6IE24, Q70EL1, Q8BL06 | |
| USP54 | |
| A synthesized peptide derived from human USP54(Accession Q70EL1), corresponding to amino acid residues E733-E783. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction