missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP51 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551667
This item is not returnable.
View return policy
Description
Immunogen sequence: LRLIYQRFVW SGTPETRKRK AKSCICHVCS THMNRLHSCL SCVFFGCFTE KHIHKHAETK QHHLAVDLYH GVIYCFMCKD YVYDKDIEQI AKETKEKILR LLTSTSTDVS HQQFMTSGFE DKQSTCETKE QEP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 83%.
USP51 (Ubiquitin Specific Peptidase 51) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include cysteine-type endopeptidase activity and thiol-dependent ubiquitinyl hydrolase activity. Specifically deubiquitinates 'Lys-14' (H2AK13Ub) and 'Lys-16'(H2AK15Ub) of histone H2A regulating the DNA damage response at double-strand breaks (DSBs). USP51 is recruited to chromatin after DNA damage and regulates the dynamic assembly/disassembly of TP53BP1 and BRCA1. Exhibits also activity for 'Lys-27' or 'Lys-63'-linked di-ubiquitin.
Specifications
| USP51 | |
| Polyclonal | |
| Unconjugated | |
| USP51 | |
| AV136873; Deubiquitinating enzyme 51; RGD1562211; Ubiquitin carboxyl-terminal hydrolase 51; ubiquitin specific peptidase 51; ubiquitin specific protease 51; ubiquitin thioesterase 51; ubiquitin thiolesterase 51; ubiquitin-specific-processing protease 51; Usp51 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158880 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q70EK9 | |
| USP51 | |
| Recombinant protein corresponding to Human USP51. Recombinant protein control fragment (Product #RP-92108). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction