missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP50 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539215
This item is not returnable.
View return policy
Description
Specifications
| USP50 | |
| Polyclonal | |
| Unconjugated | |
| USP50 | |
| USP50 | |
| A synthetic peptide derived from the C-terminal region of human USP50 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q70EL3, Q6P8X6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 373509, 75083 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction