missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP5 Mouse anti-Human, Clone: 3F12C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66213-1-IG-150UL
This item is not returnable.
View return policy
Description
USP5 (ubiquitin carboxyl-terminal hydrolase 5) cleaves linear and branched multiubiquitin polymers with a marked preference for branched polymers. It is involved in uchored Lys-48-linked polyubiquitin disassembly, and also binds linear and Lys-63 linked polyubiquitin with a lower affinity. Knock-down of USP5 causes the accumulation of p53/TP53 and an increase in p53/TP53 transcriptional activity because the uchored polyubiquitin that accumulates is able to compete with ubiquitinated p53/TP53 but not with MDM2 for proteasomal recognition.Specifications
| USP5 | |
| Monoclonal | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P45974 | |
| USP5 | |
| USP5 Fusion Protein Ag7575 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 3F12C10 | |
| Unconjugated | |
| USP5 | |
| Deubiquitinating enzyme 5, Isopeptidase T, ISOT, Ubiquitin thiolesterase 5, USP5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 8078 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction