missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598578
This item is not returnable.
View return policy
Description
USP48 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications
| USP48 | |
| Polyclonal | |
| Unconjugated | |
| USP48 | |
| 2810449C13Rik; AI115503; BC021769; D330022K21Rik; Deubiquitinating enzyme 48; Kiaa4202; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 48; RAP1GA1; Synaptic ubiquitin-specific protease; synUSP; Ubiquitin carboxyl-terminal hydrolase 48; Ubiquitin carboxyl-terminal hydrolase 48-like protein; ubiquitin specific peptidase 48; ubiquitin specific protease 31; ubiquitin specific protease 48; ubiquitin thioesterase 48; ubiquitin thiolesterase 48; Ubiquitin-specific peptidase 48; Ubiquitin-specific protease 48; Ubiquitin-specific-processing protease 48; Usp31; Usp48 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 84196 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.54 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q86UV5 | |
| USP48 | |
| Recombinant Human Ubiquitin carboxyl-terminal hydrolase 48 protein (358-508AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction