missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP48 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12076-1-AP-150UL
This item is not returnable.
View return policy
Description
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.Specifications
| USP48 | |
| Polyclonal | |
| Unconjugated | |
| USP48 | |
| Deubiquitinating enzyme 48, RAP1GA1, Ubiquitin thiolesterase 48, USP31, USP48 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170707, 362636, 84196 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3V0C5, Q76LT8, Q86UV5 | |
| Usp48 | |
| USP48 Fusion Protein Ag2749 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction