missing translation for 'onlineSavingsMsg'
Learn More

USP48 Antibody (OTI1F10) [PE], Novus Biologicals™

Product Code. 30687676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687676 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687676 Supplier Novus Biologicals Supplier No. NBP344676PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP48 Monoclonal antibody specifically detects USP48 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP48
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI1F10
Conjugate PE
Formulation PBS
Gene Alias Deubiquitinating enzyme 48, DKFZp762M1713, EC 3.1.2.15, EC 3.4.19.12, FLJ11328, FLJ20103, FLJ23054, FLJ23277, MGC132556, MGC14879, RAP1GA1, ubiquitin carboxyl-terminal hydrolase 48, ubiquitin specific peptidase 48, ubiquitin specific protease 31, ubiquitin specific protease 48, ubiquitin thioesterase 48, Ubiquitin thiolesterase 48, Ubiquitin-specific-processing protease 48, USP31
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 361-743 of human USP48 (NP_115612) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84196
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.