missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP48 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38119-100ul
This item is not returnable.
View return policy
Description
USP48 Polyclonal antibody specifically detects USP48 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| USP48 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Deubiquitinating enzyme 48, DKFZp762M1713, EC 3.1.2.15, EC 3.4.19.12, FLJ11328, FLJ20103, FLJ23054, FLJ23277, MGC132556, MGC14879, RAP1GA1, ubiquitin carboxyl-terminal hydrolase 48, ubiquitin specific peptidase 48, ubiquitin specific protease 31, ubiquitin specific protease 48, ubiquitin thioesterase 48, Ubiquitin thiolesterase 48, Ubiquitin-specific-processing protease 48, USP31 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human USP48 (NP_115612.4).,, Sequence:, MAPRLQLEKAAWRWAETVRPEEVSQEHIETAYRIWLEPCIRGVCRRNCKGNPNCLVGIGEHIWLGEIDENSFHNIDDPNCERRKKNSFVGLTNLGATCYVNTFLQVWFLNLELRQALYLCPSTCSDYMLGDGIQEEKDYEPQTICEHLQYLFALLQNSNRRYIDPSGFVKALGLDTGQQQDAQEFSKLFMSLLEDTLSKQKNPDVRNIVQQQFCGEYAYVTVCNQCGRESKLLSKFYELELNIQGHKQLTDCISEFLKEEKLEGDNRYFCENCQSKQNATRKIRLLSLPCTLNLQLMRFV | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84196 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction