missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP46 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13502-1-AP-150UL
This item is not returnable.
View return policy
Description
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.Specifications
| USP46 | |
| Polyclonal | |
| Unconjugated | |
| USP46 | |
| Deubiquitinating enzyme 46, Ubiquitin thiolesterase 46, USP46 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289584, 64854, 69727 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F1M625, P62068, P62069 | |
| Usp46 | |
| USP46 Fusion Protein Ag4330 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction