Learn More
Invitrogen™ USP46 Monoclonal Antibody (CL0364)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524601
Description
Immunogen sequence: LFDNYMQQDA HEFLNYLLNT IADILQEEKK QEKQNGKLKN GNMNEPAENN KPELTWVHEI FQGTLTNETR CLNCETVSSK DEDFLDLSVD VEQN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%. Binds to an epitope located within the peptide sequence TNETRCLNCE as determined by overlapping synthetic peptides.
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
Specifications
| USP46 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62068 | |
| USP46 | |
| Recombinant protein fragment of Human USP46 (Product #RP-89251) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| CL0364 | |
| Unconjugated | |
| USP46 | |
| 1190009E20Rik; 2410018I08Rik; AI451644; Deubiquitinating enzyme 46; RGD1564808; Ubiquitin carboxyl-terminal hydrolase 46; ubiquitin specific peptidase 46; ubiquitin specific protease 46; ubiquitin thioesterase 46; ubiquitin thiolesterase 46; ubiquitin-specific-processing protease 46; Usp46 | |
| Mouse | |
| Protein A | |
| RUO | |
| 64854 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.