missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP45 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556205
This item is not returnable.
View return policy
Description
Immunogen sequence: EKAKRSKRPT VPHDEDSSDD IAVGLTCQHV SHAISVNHVK RAIAENLWSV CSECLEERRF YDGQLVLTSD IWLCLKCGFQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
The protein encoded by this gene is a deubiquitylase that binds ERCC1, the catalytic subunit of the XPF-ERCC1 DNA repair endonuclease. This endonuclease is a critical regulator of DNA repair processes, and the deubiquitylase activity of the encoded protein is important for maintaining the DNA repair ability of XPF-ERCC1.
Specifications
| USP45 | |
| Polyclonal | |
| Unconjugated | |
| Usp45 | |
| 3110003C05Rik; 4930550B20Rik; AI843191; Deubiquitinating enzyme 45; Gcap7; granule cell antiserum positive 7; ubiquitin carboxyl-terminal hydrolase 45; ubiquitin specific peptidase 45; ubiquitin specific petidase 45; ubiquitin specific protease 45; ubiquitin thioesterase 45; ubiquitin thiolesterase 45; ubiquitin-specific-processing protease 45; USP45 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q70EL2 | |
| Usp45 | |
| Recombinant protein corresponding to Human USP45. Recombinant protein control fragment (Product #RP-94741). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction