missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP44 Antibody (OTI1D1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74811R
This item is not returnable.
View return policy
Description
USP44 Monoclonal specifically detects USP44 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| USP44 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| USP44 | |
| Recombinant protein fragment corresponding to amino acids 420-712 of human USP44 (NP_115523) produced in E.coli. | |
| 0.1 mL | |
| 84101.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1D1 | |
| Western Blot, Immunohistochemistry | |
| Deubiquitinating enzyme 44, DKFZp434D0127, EC 3.1.2.15, EC 3.4.19.12, FLJ14528, ubiquitin carboxyl-terminal hydrolase 44, ubiquitin specific peptidase 44, ubiquitin specific protease 44, ubiquitin thioesterase 44, Ubiquitin thiolesterase 44, Ubiquitin-specific-processing protease 44 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction