missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101945
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP43.
May recognize and hydrolyze the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins. [UniProt].
Specifications
| USP43 | |
| Polyclonal | |
| Unconjugated | |
| USP43 | |
| Deubiquitinating enzyme 43; ubiquitin carboxyl-terminal hydrolase 43; ubiquitin specific peptidase 43; ubiquitin specific protease 43; ubiquitin thioesterase 43; ubiquitin thiolesterase 43; ubiquitin-specific-processing protease 43; USP43 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 124739, 216835 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q70EL4, Q8BUM9 | |
| USP43 | |
| A synthesized peptide derived from human USP43(Accession Q70EL4), corresponding to amino acid residues N113-L163. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction