missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP43 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88562
This item is not returnable.
View return policy
Description
USP43 Polyclonal specifically detects USP43 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| USP43 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 124739 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| Deubiquitinating enzyme 43, EC 3.1.2.15, EC 3.4.19.12, FLJ30626, ubiquitin carboxyl-terminal hydrolase 43, ubiquitin specific peptidase 43, ubiquitin specific protease 43, ubiquitin thioesterase 43, Ubiquitin thiolesterase 43, Ubiquitin-specific-processing protease 43 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human USP43 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction