missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP42 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566720
This item is not returnable.
View return policy
Description
Immunogen sequence: TIVDKASESSD PSAYQNQPGS SEAVSPGDMD AGSASWGAVS SLNDVSNHTL SLGPVPGAVV YSSSSVPDKS KPSPQKDQAL GDGIAPPQKV L Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.
USP42 (ubiquitin specific peptidase 42) is a 1,325 amino acid protein that belongs to the peptidase C19 family. Expressed in a variety of tissues, USP42 functions to catalyze the conversion of a ubiquitin C-terminal thioester to a free ubiquitin and a thiol. The gene encoding USP42 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
Specifications
| USP42 | |
| Polyclonal | |
| Unconjugated | |
| Usp42 | |
| 2410140K03Rik; 3110031A07Rik; A630018G05Rik; D5Ertd591e; Deubiquitinating enzyme 42; ubiquitin carboxyl-terminal hydrolase 42; ubiquitin specific peptidase 42; ubiquitin specific protease 42; ubiquitin thioesterase 42; ubiquitin thiolesterase 42; ubiquitin-specific-processing protease 42; Usp42 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84132 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H9J4 | |
| Usp42 | |
| Recombinant Human USP42. Recombinant protein control fragment (Product #RP-105344). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction