missing translation for 'onlineSavingsMsg'
Learn More

USP41 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15819693
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15819693 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819693 Supplier Invitrogen Supplier No. PA571281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

USP41 Polyclonal Antibody

USP41 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen USP41
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USP41
Gene Accession No. Q3LFD5
Gene Alias deubiquitinating enzyme 41; putative ubiquitin carboxyl-terminal hydrolase 41; ubiquitin specific peptidase 41; ubiquitin specific protease 41; ubiquitin specific proteinase 41; ubiquitin thioesterase 41; ubiquitin thiolesterase 41; ubiquitin-specific-processing protease 41; USP41
Gene Symbols USP41
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human USP41.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 373856
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.