missing translation for 'onlineSavingsMsg'
Learn More

USP40 Antibody (OTI4F7) [Alexa Fluor™ 488], Novus Biologicals™

Product Code. 30689359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689359 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689359 Supplier Novus Biologicals Supplier No. NBP345031AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP40 Monoclonal antibody specifically detects USP40 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP40
Applications Western Blot
Classification Monoclonal
Clone OTI4F7
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias deubiquitinating enzyme 40, ubiquitin specific peptidase 40, ubiquitin thiolesterase 40, ubiquitin-specific-processing protease 40
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 767-912 of human USP40(NP_060688) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 55230
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.