Learn More
Invitrogen™ USP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533106
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Dog, Bat, Bovine, Elephant, Panda, Horse, Rabbit, Pig (100%); Hamster, Opossum (94%); Mouse, Rat, Turkey, Platypus (89%); Chicken, Xenopus (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
The protein encoded by this gene is a protease that deubiquitinates target proteins such as ADORA2A and TRIM21. The encoded protein shuttles between the nucleus and cytoplasm and is involved in maintaining operational fidelity in the endoplasmic reticulum. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| USP4 | |
| Polyclonal | |
| Unconjugated | |
| USP4 | |
| Deubiquitinating enzyme 4; F730026I20Rik; mKIAA4155; ubiquitin carboxyl-terminal esterase 4; ubiquitin carboxyl-terminal hydrolase 4; ubiquitin specific peptidase 4; ubiquitin specific peptidase 4 (proto-oncogene); ubiquitin specific protease 4 (proto-oncogene); ubiquitin thioesterase 4; ubiquitin thiolesterase 4; ubiquitin-specific processing protease 4; ubiquitin-specific-processing protease 4; Ubiquitous nuclear protein; ubiquitous nuclear protein homolog; Unp; Unph; USP4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100053443, 100359136, 100512072, 476624, 508042, 7375 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 0.1% sodium azide | |
| A6QR55, Q13107 | |
| USP4 | |
| Synthetic 18 amino acid peptide from internal region of human USP4. | |
| 50 μg | |
| Primary | |
| Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.