missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP39 Recombinant Rabbit Monoclonal Antibody (24GB1810)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557819
This item is not returnable.
View return policy
Description
USP39 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
USP39 may play a role in mRNA splicing. It is unsure if the protein really exhibits hydrolase activity. It could be a competitor of ubiquitin C-terminal hydrolases.
Specifications
| USP39 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| USP39 | |
| 65K; AA408960; AI894154; CGI-21; D6Wsu157e; HSPC332; Inactive ubiquitin-specific peptidase 39; PRO2855; SAD1; SAD1 homolog; small nuclear ribonucleoprotein 65kDa (U4/U6.U5); SnRNP assembly defective 1 homolog; SNRNP65; U4/U6.U5 tri-snRNP-associated 65 kDa protein; U4/U6.U5 tri-snRNP-associated protein 2; ubiquitin specific peptidase 39; ubiquitin specific protease 39; Usp39 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10713, 28035, 297336 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1810 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3TIX9, Q53GS9 | |
| USP39 | |
| A synthesized peptide derived from human USP39 (aa 201-350). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction