missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
USP39 Monoclonal antibody specifically detects USP39 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | USP39 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 9E7O2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | CGI-21, HSPC332, Inactive ubiquitin-specific peptidase 39,65K, SAD1 homolog, SAD1MGC75069, small nuclear ribonucleoprotein 65kDa (U4/U6.U5), SnRNP assembly defective 1 homolog, SNRNP65, U4/U6.U5 tri-snRNP-associated 65 kDa protein, U4/U6.U5 tri-snRNP-associated protein 2, ubiquitin specific peptidase 39, ubiquitin specific protease 39 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200-300 of human USP39 (Q53GS9). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?