missing translation for 'onlineSavingsMsg'
Learn More

USP39 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18331246 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331246 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331246 Supplier Novus Biologicals Supplier No. NBP317961100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP39 Polyclonal antibody specifically detects USP39 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP39
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias CGI-21, HSPC332, Inactive ubiquitin-specific peptidase 39,65K, SAD1 homolog, SAD1MGC75069, small nuclear ribonucleoprotein 65kDa (U4/U6.U5), SnRNP assembly defective 1 homolog, SNRNP65, U4/U6.U5 tri-snRNP-associated 65 kDa protein, U4/U6.U5 tri-snRNP-associated protein 2, ubiquitin specific peptidase 39, ubiquitin specific protease 39
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: TIVNFPITNVDLREYLSEEVQAVHKNTTYDLIANIVHDGKPSEGSYRIHVLHHGTGKWYELQDLQVTDILPQMITLSEAYIQIWKRRDNDETN
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 10713
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.