missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565745
This item is not returnable.
View return policy
Description
Immunogen sequence: EDGHLKCPRSA KPQDAVVPES SSCAPSANGW CPGDRMGLSQ APPVSWNGER ESDVVQELLK YSSDKAYGRK VLTWDGKMSA VSQDAIEDSR QARTETVVDD WDEEFDRGKE KKIKKFKREK RRNFNAFQKL QTRRNFWSVT HPAK Highest antigen sequence identity to the following orthologs - mouse 70%, rat 68%.
May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 'Lys-4' trimethylation (H3K4).
Specifications
| USP36 | |
| Polyclonal | |
| Unconjugated | |
| USP36 | |
| 2700002L06Rik; deubiquitinating enzyme 36; DUB1; Kiaa1453; mKIAA1453; Ubiquitin carboxyl-terminal hydrolase 36; ubiquitin specific peptidase 36; ubiquitin specific protease 36; ubiquitin thioesterase 36; ubiquitin thiolesterase 36; ubiquitin-specific-processing protease 36; Usp36 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57602 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P275 | |
| USP36 | |
| Recombinant Human USP36. Recombinant protein control fragment (Product #RP-106421). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction