missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
USP35 Polyclonal specifically detects USP35 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | USP35 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | Deubiquitinating enzyme 35, EC 3.4.19.12, KIAA1372, ubiquitin carboxyl-terminal hydrolase 35, ubiquitin specific peptidase 35, ubiquitin specific protease 35, ubiquitin thioesterase 35, Ubiquitin thiolesterase 35, Ubiquitin-specific-processing protease 35, USP34 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human USP35 (NP_065849). Peptide sequence MEAISKDNILYLQEQEKEARSRAAYISALPTSPHWGRGFDEDKDEDEGSP |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?