missing translation for 'onlineSavingsMsg'
Learn More

USP34 Antibody (2E2), Novus Biologicals™

Product Code. 18338658 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338658 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338658 Supplier Novus Biologicals Supplier No. H00009736M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP34 Monoclonal antibody specifically detects USP34 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP34
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2E2
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055524
Gene Alias Deubiquitinating enzyme 34, EC 3.1.2.15, EC 3.4.19.12, FLJ43910, KIAA0570MGC104459, KIAA0729ubiquitin carboxyl-terminal hydrolase 34, ubiquitin specific peptidase 34, ubiquitin specific protease 34, ubiquitin thioesterase 34, Ubiquitin thiolesterase 34, Ubiquitin-specific-processing protease 34
Host Species Mouse
Immunogen USP34 (NP_055524, 3296 a.a. ~ 3395 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CKEFKDLHCSKDSTLAEEESEFPSTSISAVLSDLADLRSCDGQALPSQDPEVALSLSCGHSRGLFSHMQQHDILDTLCRTIESTIHVVTRISGKGNQAAS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9736
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.