missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP33 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £443.00
Specifications
| Antigen | USP33 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18418330
|
Novus Biologicals
NBP1-82931-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263246
|
Novus Biologicals
NBP1-82931 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP33 Polyclonal specifically detects USP33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| USP33 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Deubiquitinating enzyme 33, EC 3.1.2.15, EC 3.4.19.12, hVDU1, KIAA1097MGC16868, pVHL-interacting deubiquitinating enzyme 1, ubiquitin carboxyl-terminal hydrolase 33, ubiquitin specific peptidase 33, ubiquitin thioesterase 33, Ubiquitin thiolesterase 33, Ubiquitin-specific-processing protease 33, VDU1ubiquitin specific protease 33, VHL-interacting deubiquitinating enzyme 1 | |
| USP33 | |
| IgG | |
| Affinity Purified | |
| Specificity of human USP33 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| GPCR, Protein Turnover, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 23032 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ESQVDHSTIHSQETKHYLTVNLTTLRVWCYACSKEVFLDRKLGTQPSLPHVRQPHQIQENSVQDFKIPSNTTLKTPLVAVFDDLDIEADEEDELRARGLTGLKNIGNTCYMNAALQALS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title