missing translation for 'onlineSavingsMsg'
Learn More

USP33 Antibody (5B5), Novus Biologicals™

Product Code. 18396558 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396558 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396558 Supplier Novus Biologicals Supplier No. H00023032M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP33 Monoclonal antibody specifically detects USP33 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP33
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 5B5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055832
Gene Alias Deubiquitinating enzyme 33, EC 3.1.2.15, EC 3.4.19.12, hVDU1, KIAA1097MGC16868, pVHL-interacting deubiquitinating enzyme 1, ubiquitin carboxyl-terminal hydrolase 33, ubiquitin specific peptidase 33, ubiquitin thioesterase 33, Ubiquitin thiolesterase 33, Ubiquitin-specific-processing protease 33, VDU1ubiquitin specific protease 33, VHL-interacting deubiquitinating enzyme 1
Host Species Mouse
Immunogen USP33 (NP_055832, 327 a.a. ~ 426 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SDRAENENGSRCFSEDNNETTMLIQDDENNSEMSKDWQKEKMCNKINKVNSEGEFDKDRDSISETVDLNNQETVKVQIHSRASEYITDVHSNDLSTPQIL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Turnover, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 23032
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.