missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP32 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13509
This item is not returnable.
View return policy
Description
USP32 Polyclonal specifically detects USP32 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| USP32 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Deubiquitinating enzyme 32, EC 3.1.2.15, EC 3.4.19.12, NY-REN-60, Renal carcinoma antigen NY-REN-60, ubiquitin carboxyl-terminal hydrolase 32, ubiquitin specific peptidase 32, ubiquitin specific protease 32, ubiquitin thioesterase 32, Ubiquitin thiolesterase 32, USP10Ubiquitin-specific-processing protease 32 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84669 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| USP32 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: VVIEPSSVLNGGKYSFGTAAHPMEQVEDRIGSSLSYVNTTEEKFSDNISTASEASETAGSGFLYSATPGADVCFARQHNTSDNNNQCLLGANGNILLHLNPQKPGAIDNQPLVTQEPVKATSLTLEGGRLKRTPQLIHGRDYEMVPE | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction