missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104199
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP31.
May recognize and hydrolyze the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications
| USP31 | |
| Polyclonal | |
| Unconjugated | |
| USP31 | |
| 6330567E21Rik; Deubiquitinating enzyme 31; KIAA1203; mKIAA1203; ubiquitin carboxyl-terminal hydrolase 31; ubiquitin specific peptidase 31; ubiquitin specific protease 31; ubiquitin specific proteinase 31; ubiquitin thioesterase 31; ubiquitin thiolesterase 31; ubiquitin-specific-processing protease 31; USP31 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 308959, 57478, 76179 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q70CQ4 | |
| USP31 | |
| A synthesized peptide derived from human USP31(Accession Q70CQ4), corresponding to amino acid residues S225-S275. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction