missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP30 Antibody (CL4438) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59049IR
This item is not returnable.
View return policy
Description
USP30 Monoclonal antibody specifically detects USP30 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| USP30 | |
| Monoclonal | |
| DyLight 755 | |
| Deubiquitinating enzyme 30, EC 3.1.2.15, EC 3.4.19.12, FLJ40511, MGC10702, ubiquitin carboxyl-terminal hydrolase 30, ubiquitin specific peptidase 30, ubiquitin specific protease 30, ubiquitin thioesterase 30, Ubiquitin thiolesterase 30, ubiquitin-specific protease 30, Ubiquitin-specific-processing protease 30, Ub-specific protease 30 | |
| This antibody was developed using a recombinant protein derived from Q70CQ3, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 84749 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL4438 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction