missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP30 Antibody (CL4438) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44104
This item is not returnable.
View return policy
Description
USP30 Monoclonal antibody specifically detects USP30 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
| USP30 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Deubiquitinating enzyme 30, EC 3.1.2.15, EC 3.4.19.12, FLJ40511, MGC10702, ubiquitin carboxyl-terminal hydrolase 30, ubiquitin specific peptidase 30, ubiquitin specific protease 30, ubiquitin thioesterase 30, Ubiquitin thiolesterase 30, ubiquitin-specific protease 30, Ubiquitin-specific-processing protease 30, Ub-specific protease 30 | |
| This antibody was generated using a recombinant protein sequence of Q70CQ3, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 84749 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL4438 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction